missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human NETO2 Full-length ORF (AAH12381.1, 1 a.a. - 148 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00081831-P01.10ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16105253

  • 335.00€ / 10 microgramos
Fecha estimada de envío: 05-06-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This gene encodes a predicted transmembrane protein containing two extracellular CUB domains followed by a low-density lipoprotein class A (LDLa) domain. It also has an intracellular FXNPXY-like motif, which has been shown in other proteins to be essential for the internalization of clathrin coated pits during endocytosis. Alternatively spliced transcript variants have been observed, but they have not been fully characterized. [provided by RefSeq]

Sequence: MACKTAFNKTGFQEVFDPPHYELFSLRDKEISADLADLSEELDNYQKMRRSSTASRCIHDHHCGSQASSVKQSRTNLSSMELPFRNDFAQPQPMKTFNSTFKKSSYTFKQGHECPEQALEDRVMEEIPCEIYVRGREDSAQASISIDF
Especificaciones

Especificaciones

AAH12381.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
43.4kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ10430/FLJ14724/FLJ90456/NEOT2
NETO2
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
81831
NETO2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MACKTAFNKTGFQEVFDPPHYELFSLRDKEISADLADLSEELDNYQKMRRSSTASRCIHDHHCGSQASSVKQSRTNLSSMELPFRNDFAQPQPMKTFNSTFKKSSYTFKQGHECPEQALEDRVMEEIPCEIYVRGREDSAQASISIDF
RUO
NETO2
Wheat Germ (in vitro)
GST
Liquid
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales