missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human NEK3 Full-length ORF (NP_002489.1, 1 a.a. - 506 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00004752-P01.10ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16100678

  • 335.00€ / 10 microgramos
Fecha estimada de envío: 07-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This gene encodes a member of the NimA (never in mitosis A) family of serine/threonine protein kinases. The encoded protein differs from other NimA family members in that it is not cell cycle regulated and is found primarily in the cytoplasm. The kinase is activated by prolactin stimulation, leading to phosphorylation of VAV2 guanine nucleotide exchange factor, paxillin, and activation of the RAC1 GTPase. Multiple transcript variants encoding different isoforms have been found for this gene

Sequence: MDDYMVLRMIGEGSFGRALLVQHESSNQMFAMKEIRLPKSFSNTQNSRKEAVLLAKMKHPNIVAFKESFEAEGHLYIVMEYCDGGDLMQKIKQQKGKLFPEDMILNWFTQMCLGVNHIHKKRVLHRDIKSKNIFLTQNGKVKLGDFGSARLLSNPMAFACTYVGTPYYVPPEIWENLPYNNKSDIWSLGCILYELCTLKHPFQANSWKNLILKVCQGCISPLPSHYSYELQFLVKQMFKRNPSHRPSATTLLSRGIVARLVQKCLPPEIIMEYGEEVLEEIKNSKHNTPRKKTNPSRIRIALGNEASTVQEEEQDRKGSHTDLESINENLVESALRRVNREEKGNKSVHLRKASSPNLHRRQWEKNVPNTALTALENASILTSSLTAEDDRGGSVIKYSKNTTRKQWLKETPDTLLNILKNADLSLAFQTYTIYRPGSEGFLKGPLSEETEASDSVDGGHDSVILDPERLEPGLDEEDTDFEEEDDNPDWVSELKKRAGWQGLCDR
Especificaciones

Especificaciones

NP_002489.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
84.1kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HSPK36/MGC29949
NEK3
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
4752
NEK3 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MDDYMVLRMIGEGSFGRALLVQHESSNQMFAMKEIRLPKSFSNTQNSRKEAVLLAKMKHPNIVAFKESFEAEGHLYIVMEYCDGGDLMQKIKQQKGKLFPEDMILNWFTQMCLGVNHIHKKRVLHRDIKSKNIFLTQNGKVKLGDFGSARLLSNPMAFACTYVGTPYYVPPEIWENLPYNNKSDIWSLGCILYELCTLKHPFQANSWKNLILKVCQGCISPLPSHYSYELQFLVKQMFKRNPSHRPSATTLLSRGIVARLVQKCLPPEIIMEYGEEVLEEIKNSKHNTPRKKTNPSRIRIALGNEASTVQEEEQDRKGSHTDLESINENLVESALRRVNREEKGNKSVHLRKASSPNLHRRQWEKNVPNTALTALENASILTSSLTAEDDRGGSVIKYSKNTTRKQWLKETPDTLLNILKNADLSLAFQTYTIYRPGSEGFLKGPLSEETEASDSVDGGHDSVILDPERLEPGLDEEDTDFEEEDDNPDWVSELKKRAGWQGLCDR
RUO
NEK3
Wheat Germ (in vitro)
GST
Liquid
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales