missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human NDUFA9 Full-length ORF (NP_004993.1, 1 a.a. - 377 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00004704-P01.10ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16190588

  • 310.00€ / 10 microgramos


Fecha estimada de envío
24-02-2021
Añadir a la cesta

Descripción

Descripción

Sequence: MAAAAQSRVVRVLSMSRSAITAIATSVCHGPPCRQLHHALMPHGKGGRSSVSGIVATVFGATGFLGRYVVNHLGRMGSQVIIPYRCDKYDIMHLRPMGDLGQLLFLEWDARDKDSIRRVVQHSNVVINLIGRDWETKNFDFEDVFVKIPQAIAQLSKEAGVEKFIHVSHLNANIKSSSRYLRNKAVGEKVVRDAFPEAIIVKPSDIFGREDRFLNSFASMHRFGPIPLGSLGWKTVKQPVYVVDVSKGIVNAVKDPDANGKSFAFVGPSRYLLFHLVKYIFAVAHRLFLPFPLPLFAYRWVARVFEISPFEPWITRDKVERMHITDMKLPHLPGLEDLGIQATPLELKAIEVLRRHRTYRWLSAEIEDVKPAKTVNI
Especificaciones

Especificaciones

NP_004993.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
68.9kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC111043|NDUFS2L|SDR22E1
NDUFA9
Yes
wheat germ expression system
Liquid
4704
NDUFA9 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAAAAQSRVVRVLSMSRSAITAIATSVCHGPPCRQLHHALMPHGKGGRSSVSGIVATVFGATGFLGRYVVNHLGRMGSQVIIPYRCDKYDIMHLRPMGDLGQLLFLEWDARDKDSIRRVVQHSNVVINLIGRDWETKNFDFEDVFVKIPQAIAQLSKEAGVEKFIHVSHLNANIKSSSRYLRNKAVGEKVVRDAFPEAIIVKPSDIFGREDRFLNSFASMHRFGPIPLGSLGWKTVKQPVYVVDVSKGIVNAVKDPDANGKSFAFVGPSRYLLFHLVKYIFAVAHRLFLPFPLPLFAYRWVARVFEISPFEPWITRDKVERMHITDMKLPHLPGLEDLGIQATPLELKAIEVLRRHRTYRWLSAEIEDVKPAKTVNI
RUO
NDUFA9
Wheat Germ (in vitro)
GST
Videos
SDS
Documentos