Learn More
Abnova™ Human NCF4 Partial ORF (AAH02798, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00004689-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
The protein encoded by this gene is a cytosolic regulatory component of the superoxide-producing phagocyte NADPH-oxidase, a multicomponent enzyme system important for host defense. This protein is preferentially expressed in cells of myeloid lineage. It interacts primarily with neutrophil cytosolic factor 2 (NCF2/p67-phox) to form a complex with neutrophil cytosolic factor 1 (NCF1/p47-phox), which further interacts with the small G protein RAC1 and translocates to the membrane upon cell stimulation. This complex then activates flavocytochrome b, the membrane-integrated catalytic core of the enzyme system. The PX domain of this protein can bind phospholipid products of the PI(3) kinase, which suggests its role in PI(3) kinase-mediated signaling events. The phosphorylation of this protein was found to negatively regulate the enzyme activity. Alternatively spliced transcript variants encoding distinct isoforms have been observed. [provided by RefSeq]
Sequence: MAVAQQLRAESDFEQLPDDVAISANIADIEEKRGFTSHFVFVIEVKTKGGSKYLIYRRYRQFHALQSKLEERFGPDSKSSALACTLPTLPAKVYVGVKQEEspecificaciones
AAH02798 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAVAQQLRAESDFEQLPDDVAISANIADIEEKRGFTSHFVFVIEVKTKGGSKYLIYRRYRQFHALQSKLEERFGPDSKSSALACTLPTLPAKVYVGVKQE | |
RUO | |
NCF4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
4689 | |
NCF4 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC3810/NCF/P40PHOX/SH3PXD4 | |
NCF4 | |
Recombinant | |
wheat germ expression system |