missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human NANOS3 Partial ORF (XP_292819.2, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00342977-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: MGTFDLWTDYLGLAHLVRALSGKEGPETRLSPQPEPEPMLEPVSALEPMPAPESVPVPGPKDQKRSLESSPAPERLCSFCKHNGESRAIYQSHVLKDEAGEspecificaciones
XP_292819.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MGTFDLWTDYLGLAHLVRALSGKEGPETRLSPQPEPEPMLEPVSALEPMPAPESVPVPGPKDQKRSLESSPAPERLCSFCKHNGESRAIYQSHVLKDEAG | |
RUO | |
NANOS3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
342977 | |
NANOS3 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC120114/NANOS1L/NOS3 | |
NANOS3 | |
Recombinant | |
wheat germ expression system |