missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MUC4 Partial ORF (NP_004523, 79 a.a. - 188 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00004585-Q01.25ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16171465

  • 528.00€ / 25 microgramos
Fecha estimada de envío: 31-10-2023
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

The major constituents of mucus, the viscous secretion that covers epithelial surfaces such as those in the trachea, colon, and cervix, are highly glycosylated proteins called mucins. These glycoproteins play important roles in the protection of the epithelial cells and have been implicated in epithelial renewal and differentiation. This gene encodes an integral membrane glycoprotein found on the cell surface, although secreted isoforms may exist. At least two dozen transcript variants of this gene have been found, although for many of them the full-length transcript has not been determined or they are found only in tumor tissues. This gene contains a region in the coding sequence which has a variable number (>100) of 48 nt tandem repeats. [provided by RefSeq]

Sequence: GVSLFPYGADAGDLEFVRRTVDFTSPLFKPATGFPLGSSLRDSLYFTDNGQIIFPESDYQIFSYPNPLPTGFTGRDPVALVAPFWDDADFSTGRGTTFYQEYETFYGEHS
Especificaciones

Especificaciones

NP_004523
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
GVSLFPYGADAGDLEFVRRTVDFTSPLFKPATGFPLGSSLRDSLYFTDNGQIIFPESDYQIFSYPNPLPTGFTGRDPVALVAPFWDDADFSTGRGTTFYQEYETFYGEHS
RUO
MUC4
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
4585
MUC4 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HSA276359
MUC4
Recombinant
wheat germ expression system
Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales