missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MTR Partial ORF (NP_000245, 1094 a.a. - 1203 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00004548-Q01.10ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16108021

  • 341.70€ / 10 microgramos
Envío estimado: 21-01-2022

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

MTR encodes the enzyme 5-methyltetrahydrofolate-homocysteine methyltransferase. This enzyme, also known as cobalamin-dependent methionine synthase, catalyzes the final step in methionine biosynthesis. Mutations in MTR have been identified as the underlying cause of methylcobalamin deficiency complementation group G. [provided by RefSeq]

Sequence: RDYLGLFAVACFGVEELSKAYEDDGDDYSSIMVKALGDRLAEAFAEELHERVRRELWAYCGSEQLDVADLRRLRYKGIRPAPGYPSQPDHTEKLTMWRLADIEQSTGIRL
Especificaciones

Especificaciones

NP_000245
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
RDYLGLFAVACFGVEELSKAYEDDGDDYSSIMVKALGDRLAEAFAEELHERVRRELWAYCGSEQLDVADLRRLRYKGIRPAPGYPSQPDHTEKLTMWRLADIEQSTGIRL
RUO
MTR
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
4548
MTR (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ33168/FLJ43216/FLJ45386/MS
MTR
Yes
Videos
SDS
Documentos
Certificados
Ofertas especiales

Ofertas especiales