Learn More
Abnova™ Human MLX Partial ORF (NP_733752.1, 51 a.a. - 150 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00006945-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
The product of this gene belongs to the family of basic helix-loop-helix leucine zipper (bHLH-Zip) transcription factors. These factors form heterodimers with Mad proteins and play a role in proliferation, determination and differentiation. This gene product may act to diversify Mad family function by its restricted association with a subset of the Mad family of transcriptional repressors, namely, Mad1 and Mad4. Alternatively spliced transcript variants encoding different isoforms have been identified for this gene. [provided by RefSeq]
Sequence: LSVPRGCREDSSHPACAKVEYAYSDNSLDPGLFVESTRKGSVVSRANSIGSTSASSVPNTDDEDSDYHQEAYKESYKDRRRRAHTQAEQKRRDAIKRGYDEspecificaciones
NP_733752.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
LSVPRGCREDSSHPACAKVEYAYSDNSLDPGLFVESTRKGSVVSRANSIGSTSASSVPNTDDEDSDYHQEAYKESYKDRRRRAHTQAEQKRRDAIKRGYD | |
RUO | |
MLX | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
6945 | |
MLX (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MAD7/MXD7/TCFL4/bHLHd13 | |
MLX | |
Recombinant | |
wheat germ expression system |