missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human MFAP1 Partial ORF (NP_005917.2, 341 a.a. - 438 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00004236-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: YYHRGAFFMDEDEEVYKRDFSAPTLEDHFNKTILPKVMQVKNFGRSGRTKYTHLVDQDTTSFDSAWGQESAQNTKFFKQKAAGVRDVFERPSAKKRKTEspecificaciones
NP_005917.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.52kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
YYHRGAFFMDEDEEVYKRDFSAPTLEDHFNKTILPKVMQVKNFGRSGRTKYTHLVDQDTTSFDSAWGQESAQNTKFFKQKAAGVRDVFERPSAKKRKT | |
RUO | |
MFAP1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
4236 | |
MFAP1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MFAP1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |