missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MECP2 Partial ORF (AAH11612, 81 a.a. - 170 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00004204-Q01.10ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16197711

  • 302.00€ / 10 microgramos


Fecha estimada de envío
16-10-2020
Añadir a la cesta

Descripción

Descripción

Sequence: PKQRRSIIRDRGPMYDDPTLPEGWTRKLKQRKSGRSAGKYDVYLINPQGKAFRSKVELIAYFEKVGDTSLDPNDFDFTVTGRGSPSRREQ
Especificaciones

Especificaciones

AAH11612
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
35.53kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
PKQRRSIIRDRGPMYDDPTLPEGWTRKLKQRKSGRSAGKYDVYLINPQGKAFRSKVELIAYFEKVGDTSLDPNDFDFTVTGRGSPSRREQ
RUO
MECP2
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
4204
MECP2 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
AUTSX3|DKFZp686A24160|MRX16|MRX79|MRXS13|MRXSL|PPMX|RTS|RTT
MECP2
Yes
Videos
Documentos