Learn More
Abnova™ Human MAP1D Full-length ORF (NP_954697.1, 1 a.a. - 335 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00254042-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
The N-terminal methionine excision pathway is an essential process in which the N-terminal methionine is removed from many proteins, thus facilitating subsequent protein modification. In mitochondria, enzymes that catalyze this reaction are celled methionine aminopeptidases (MetAps, or MAPs; EC 3.4.11.18) (Serero et al., 2003 [PubMed 14532271]).[supplied by OMIM]
Sequence: MAAPSGVHLLVRRGSHRIFSSPLNHIYLHKQSSSQQRRNFFFRRQRDISHSIVLPAAVSSAHPVPKHIKKPDYVTTGIVPDWGDSIEVKNEDQIQGLHQACQLARHVLLLAGKSLKVDMTTEEIDALVHREIISHNAYPSPLGYGGFPKSVCTSVNNVLCHGIPDSRPLQDGDIINIDVTVYYNGYHGDTSETFLVGNVDECGKKLVEVARRCRDEAIAACRAGAPFSVIGNTISHITHQNGFQVCPHFVGHGIGSYFHGHPEIWHHANDSDLPMEEGMAFTIEPIITEGSPEFKVLEDAWTVVSLDNQRSAQFEHTVLITSRGAQILTKLPHEAEspecificaciones
NP_954697.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
63.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MAP1D | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
254042 | |
MAP1D (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAAPSGVHLLVRRGSHRIFSSPLNHIYLHKQSSSQQRRNFFFRRQRDISHSIVLPAAVSSAHPVPKHIKKPDYVTTGIVPDWGDSIEVKNEDQIQGLHQACQLARHVLLLAGKSLKVDMTTEEIDALVHREIISHNAYPSPLGYGGFPKSVCTSVNNVLCHGIPDSRPLQDGDIINIDVTVYYNGYHGDTSETFLVGNVDECGKKLVEVARRCRDEAIAACRAGAPFSVIGNTISHITHQNGFQVCPHFVGHGIGSYFHGHPEIWHHANDSDLPMEEGMAFTIEPIITEGSPEFKVLEDAWTVVSLDNQRSAQFEHTVLITSRGAQILTKLPHEA | |
RUO | |
MAP1D | |
Recombinant | |
wheat germ expression system |