Learn More
ACROBiosystems Human MAG / Siglec-4a Protein, Fc Tag
Marca: ACROBiosystems MAG-H5254-100ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Myelin-associated glycoprotein (MAG), a nervous system cell adhesion molecule, is an I-type lectin that binds to sialylated glycoconjugates, including gangliosides bearing characteristic structural determinants. Preferentially binds to alpha-2,3-linked sialic acid. Binds ganglioside Gt1b. Adhesion molecule that mediates interactions between myelinating cells and neurons by binding to neuronal sialic acid-containing gangliosides and to the glycoproteins RTN4R and RTN4RL2. Protection against apoptosis is probably mediated via interaction with neuronal RTN4R and RTN4RL2. In dorsal root ganglion neurons the inhibition is mediated primarily via binding to neuronal RTN4R or RTN4RL2 and to a lesser degree via binding to neuronal gangliosides. In cerebellar granule cells the inhibition is mediated primarily via binding to neuronal gangliosides. In sensory neurons, inhibition of neurite extension depends only partially on RTN4R, RTN4RL2 and gangliosides.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 81.1 kDa. The protein migrates as 100-115 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Especificaciones
NP_002352.1 | |
100 μg | |
Gly 20 - Pro 516 | |
MAG,Siglec-4 A,GMA,S-MAG | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
81.1 kDa | |
Human MAG, Fc Tag (MAG-H5254) is expressed from human 293 cells (HEK293). It contains AA Gly 20 - Pro 516 (Accession # P20916-1). | |
-20°C | |
MAG / Siglec-4 A | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |