Learn More
Abnova™ Human LY9 Full-length ORF (NP_001028839.1, 1 a.a. - 193 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00004063-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
LY9 belongs to the SLAM family of immunomodulatory receptors (see SLAMF1; MIM 603492) and interacts with the adaptor molecule SAP (SH2D1A; MIM 300490) (Graham et al., 2006 [PubMed 16365421]).[supplied by OMIM]
Sequence: MVAPKSHTDDWAPGPFSSKPQRSQLQIFSSVLQTSLLFLLMGLRASGKDSAPTVVSGILGGSVTLPLNISVDTEIENVIWIGPKNALAFARPKENVTIMVKSYLGRLDITKWSYSLCISNLTLNDAGSYKAQINQRNFEVTTEEEFTLFVYAPFIEKLSVHVIEGDHRTLLEGSGLESIISTLAEPRVSVREGEspecificaciones
NP_001028839.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
47.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CD229/SLAMF3/hly9/mLY9 | |
LY9 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
4063 | |
LY9 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MVAPKSHTDDWAPGPFSSKPQRSQLQIFSSVLQTSLLFLLMGLRASGKDSAPTVVSGILGGSVTLPLNISVDTEIENVIWIGPKNALAFARPKENVTIMVKSYLGRLDITKWSYSLCISNLTLNDAGSYKAQINQRNFEVTTEEEFTLFVYAPFIEKLSVHVIEGDHRTLLEGSGLESIISTLAEPRVSVREG | |
RUO | |
LY9 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |