missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human LSM4 Partial ORF (NP_036453.1, 1 a.a. - 73 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00025804-Q01.10ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16195106

  • 335.00€ / 10 microgramos
Fecha estimada de envío: 10-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Sm-like proteins were identified in a variety of organisms based on sequence homology with the Sm protein family (see SNRPD2; MIM 601061). Sm-like proteins contain the Sm sequence motif, which consists of 2 regions separated by a linker of variable length that folds as a loop. The Sm-like proteins are thought to form a stable heteromer present in tri-snRNP particles, which are important for pre-mRNA splicing.[supplied by OMIM]

Sequence: MLPLSLLKTAQNHPMLVELKNGETYNGHLVSCDNWMNINLREVICTSRDGDKFWRMPECYIRGSTIKYLRIPD
Especificaciones

Especificaciones

NP_036453.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
33.77kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MLPLSLLKTAQNHPMLVELKNGETYNGHLVSCDNWMNINLREVICTSRDGDKFWRMPECYIRGSTIKYLRIPD
RUO
LSM4
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
25804
LSM4 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
YER112W
LSM4
Recombinant
wheat germ expression system
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales