Learn More
Abnova™ Human LILRA2 Full-length ORF (AAH27916.1, 1 a.a. - 466 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00011027-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Leukocyte Ig-like receptors (LIRs) are a family of immunoreceptors expressed predominantly on monocytes and B cells and at lower levels on dendritic cells and natural killer (NK) cells. All LIRs in subfamily B have an inhibitory function (see, e.g., LILRB1, MIM 604811). LIRs in subfamily A, with short cytoplasmic domains lacking an immunoreceptor tyrosine-based inhibitory motif (ITIM) and with transmembrane regions containing a charged arginine residue, may initiate stimulatory cascades. One member of subfamily A (LILRA3; MIM 604818) lacks a transmembrane region and is presumed to be a soluble receptor.[supplied by OMIM]
Sequence: MTPILTVLICLGLSLGPRTHVQAGHLPKPTLWAEPGSVIIQGSPVTLRCQGSLQAEEYHLYRENKSASWVRRIQEPGKNGQFPIPSIAREHAGRYHCQYYSHNHSSEYSDPLELVVTGAYGKPTLSALPSPVVTLGGNVTLQCVSQVAFDGFILCKEGEDEHPQRLNSHSHARGWSWAIFSVGPVSPSRRWSYRCYAYDSNSPYVWSLPSDLLELLVPGVSKKPSLSVQPGPMVAPGESLTLQCVSDVGYDRFVLYKEGERDFLQRPGWQPQAGLSQANFTLGPVSPSHGGQYRCYSAHNLSSEWSAPSDPLDILITGQFYDRPSLSVQPVPTVAPGKNVTLLCQSRGQFHTFLLTKEGAGHPPLHLRSEHQAQQNQAEFRMGPVTSAHVGTYRCYSSLSSNPYLLSLPSDPLELVVSASLGQHPQDYTVENLIRMGVAGLVLVVLGILLFEAQHSQRSLQDAAGREspecificaciones
AAH27916.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
77kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CD85H/ILT1/LIR-7/LIR7 | |
LILRA2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
11027 | |
LILRA2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
MTPILTVLICLGLSLGPRTHVQAGHLPKPTLWAEPGSVIIQGSPVTLRCQGSLQAEEYHLYRENKSASWVRRIQEPGKNGQFPIPSIAREHAGRYHCQYYSHNHSSEYSDPLELVVTGAYGKPTLSALPSPVVTLGGNVTLQCVSQVAFDGFILCKEGEDEHPQRLNSHSHARGWSWAIFSVGPVSPSRRWSYRCYAYDSNSPYVWSLPSDLLELLVPGVSKKPSLSVQPGPMVAPGESLTLQCVSDVGYDRFVLYKEGERDFLQRPGWQPQAGLSQANFTLGPVSPSHGGQYRCYSAHNLSSEWSAPSDPLDILITGQFYDRPSLSVQPVPTVAPGKNVTLLCQSRGQFHTFLLTKEGAGHPPLHLRSEHQAQQNQAEFRMGPVTSAHVGTYRCYSSLSSNPYLLSLPSDPLELVVSASLGQHPQDYTVENLIRMGVAGLVLVVLGILLFEAQHSQRSLQDAAGR | |
RUO | |
LILRA2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |