missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human LILRA2 Full-length ORF (AAH27916.1, 1 a.a. - 466 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00011027-P01.25ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16106042

  • 508.00€ / 25 microgramos
Fecha estimada de envío: 10-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Leukocyte Ig-like receptors (LIRs) are a family of immunoreceptors expressed predominantly on monocytes and B cells and at lower levels on dendritic cells and natural killer (NK) cells. All LIRs in subfamily B have an inhibitory function (see, e.g., LILRB1, MIM 604811). LIRs in subfamily A, with short cytoplasmic domains lacking an immunoreceptor tyrosine-based inhibitory motif (ITIM) and with transmembrane regions containing a charged arginine residue, may initiate stimulatory cascades. One member of subfamily A (LILRA3; MIM 604818) lacks a transmembrane region and is presumed to be a soluble receptor.[supplied by OMIM]

Sequence: MTPILTVLICLGLSLGPRTHVQAGHLPKPTLWAEPGSVIIQGSPVTLRCQGSLQAEEYHLYRENKSASWVRRIQEPGKNGQFPIPSIAREHAGRYHCQYYSHNHSSEYSDPLELVVTGAYGKPTLSALPSPVVTLGGNVTLQCVSQVAFDGFILCKEGEDEHPQRLNSHSHARGWSWAIFSVGPVSPSRRWSYRCYAYDSNSPYVWSLPSDLLELLVPGVSKKPSLSVQPGPMVAPGESLTLQCVSDVGYDRFVLYKEGERDFLQRPGWQPQAGLSQANFTLGPVSPSHGGQYRCYSAHNLSSEWSAPSDPLDILITGQFYDRPSLSVQPVPTVAPGKNVTLLCQSRGQFHTFLLTKEGAGHPPLHLRSEHQAQQNQAEFRMGPVTSAHVGTYRCYSSLSSNPYLLSLPSDPLELVVSASLGQHPQDYTVENLIRMGVAGLVLVVLGILLFEAQHSQRSLQDAAGR
Especificaciones

Especificaciones

AAH27916.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
77kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CD85H/ILT1/LIR-7/LIR7
LILRA2
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
11027
LILRA2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue
MTPILTVLICLGLSLGPRTHVQAGHLPKPTLWAEPGSVIIQGSPVTLRCQGSLQAEEYHLYRENKSASWVRRIQEPGKNGQFPIPSIAREHAGRYHCQYYSHNHSSEYSDPLELVVTGAYGKPTLSALPSPVVTLGGNVTLQCVSQVAFDGFILCKEGEDEHPQRLNSHSHARGWSWAIFSVGPVSPSRRWSYRCYAYDSNSPYVWSLPSDLLELLVPGVSKKPSLSVQPGPMVAPGESLTLQCVSDVGYDRFVLYKEGERDFLQRPGWQPQAGLSQANFTLGPVSPSHGGQYRCYSAHNLSSEWSAPSDPLDILITGQFYDRPSLSVQPVPTVAPGKNVTLLCQSRGQFHTFLLTKEGAGHPPLHLRSEHQAQQNQAEFRMGPVTSAHVGTYRCYSSLSSNPYLLSLPSDPLELVVSASLGQHPQDYTVENLIRMGVAGLVLVVLGILLFEAQHSQRSLQDAAGR
RUO
LILRA2
Wheat Germ (in vitro)
GST
Liquid
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales