Learn More
Abnova™ Human LGALS14 Full-length ORF (NP_982297.1, 1 a.a. - 168 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00056891-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene is predominantly expressed in placenta. The encoded protein belongs to the galectin (galaptin/S-lectin) family. The members of galectin family contain one or two carbohydrate recognition domains, which can bind beta-galactoside. Two alternatively spliced transcript variants encoding distinct isoforms have been observed. [provided by RefSeq]
Sequence: MSLTHRLHLCKYWGCAVSNVCRFWEGRPLPLMIVVPYTLPVSLPVGSCVIITGTPILTFVKDPQLEVNFYTGMDEDSDIAFQFRLHFGHPAIMNSCVFGIWRYEEKCYYLPFEDGKPFELCIYVRHKEYKVMVNGQRIYNFAHRFPPASVKMLQVFRDISLTRVLISDEspecificaciones
NP_982297.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
46kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CLC2/MGC22235/PPL13 | |
LGALS14 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
56891 | |
LGALS14 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSLTHRLHLCKYWGCAVSNVCRFWEGRPLPLMIVVPYTLPVSLPVGSCVIITGTPILTFVKDPQLEVNFYTGMDEDSDIAFQFRLHFGHPAIMNSCVFGIWRYEEKCYYLPFEDGKPFELCIYVRHKEYKVMVNGQRIYNFAHRFPPASVKMLQVFRDISLTRVLISD | |
RUO | |
LGALS14 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |