missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human LCORL Full-length ORF (NP_710153.2, 1 a.a. - 318 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00254251-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: MDKGRERMAAAAAAAAAAAAAAQCRSPRCAAERRGFRRELDSWRHRLMHCVGFESILEGLYGPRLRRDLSLFEDCEPEELTDWSMDEKCSFCNLQREAVSDCIPSLDSSQSTPTEELSSQGQSNTDKIECQAENYLNALFRKKDLPQNCDPNIPLVAQELMKKMIRQFAIEYISKSGKTQENRNGSIGPSIVCKSIQMNQAENSLQEEQEGPLDLTVNRMQEQNTQQGDGVLDLSTKKTSIKSEESSICDPSSENSVAGMLQVKTDEKLNVSDENTASCPLSPIKMCLNRPIEWNLNLTTASLTSCTVHNQNLKSEEKEspecificaciones
NP_710153.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
61.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ30696/MLR1/STQTL13 | |
LCORL | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
254251 | |
LCORL (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MDKGRERMAAAAAAAAAAAAAAQCRSPRCAAERRGFRRELDSWRHRLMHCVGFESILEGLYGPRLRRDLSLFEDCEPEELTDWSMDEKCSFCNLQREAVSDCIPSLDSSQSTPTEELSSQGQSNTDKIECQAENYLNALFRKKDLPQNCDPNIPLVAQELMKKMIRQFAIEYISKSGKTQENRNGSIGPSIVCKSIQMNQAENSLQEEQEGPLDLTVNRMQEQNTQQGDGVLDLSTKKTSIKSEESSICDPSSENSVAGMLQVKTDEKLNVSDENTASCPLSPIKMCLNRPIEWNLNLTTASLTSCTVHNQNLKSEEK | |
RUO | |
LCORL | |
Wheat Germ (in vitro) | |
GST | |
Liquid |