Learn More
Abnova™ Human L3MBTL Full-length ORF (AAH39820, 1 a.a. - 772 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00026013-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes the homolog of a protein identified in Drosophila as a suppressor of malignant transformation of neuroblasts and ganglion-mother cells in the optic centers of the brain. This gene product is localized to condensed chromosomes in mitotic cells. Overexpression of this gene in a glioma cell line results in improper nuclear segregation and cytokinesis producing multinucleated cells. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq]
Sequence: MRRREGHGTDSEMGQGPVRESQSSDPPALQFRISEYKPLNMAGVEQPPTPELRQEGVTEYEDGGAPAGDGEAGPQQAEDHPQNPPEDPNQDPPEDDSTCQCQACGPHQAAGPDLGSSNDGCPQLFQERSVIVENSSGSTSASELLKPMKKRKRREYQSPSEEESEPEAMEKQEEGKDPEGQPTASTPESEEWSSSQPATGEKKECWSWESYLEEQKAITAPVSLFQDSQAVTHNKNGFKLGMKLEGIDPQHPSMYFILTVAEVCGYRLRLHFDGYSECHDFWVNANSPDIHPAGWFEKTGHKLQPPKGYKEEEFSWSQYLRSTRAQAAPKHLFVSQSHSPPPLGFQVGMKLEAVDRMNPSLVCVASVTDVVDSRFLVHFDNWDDTYDYWCDPSSPYIHPVGWCQKQGKPLTPPQDYPDPDNFCWEKYLEETGASAVPTWAFKVRPPHSFLVNMKLEAVDRRNPALIRVASVEDVEDHRIKIHFDGWSHGYDFWIDADHPDIHPAGWCSKTGHPLQPPLGPREPSSASPGGCPPLSYRSLPHTRTSKYSFHHRKCPTPGCDGSGHVTGKFTAHHCLSGCPLAERNQSRLKAELSDSEASARKKNLSGFSPRKKPRHHGRIGRPPKYRKIPQEDFQTLTPDVVHQSLFMSALSAHPDRSLSVCWEQHCKLLPGVAGISASTVAKWTIDEVFGFVQTLTGCEDQARLFKDEARIVRVTHVSGKTLVWTVAQLGDLVCSDHLQEGKGILETGVHSLLCSLPTHLLAKLSFASDSQYEspecificaciones
AAH39820 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
110.66kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp586P1522/FLJ41181/H-L(3)MBT/KIAA0681/L3MBTL1/dJ138B7.3 | |
L3MBTL | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
26013 | |
L3MBTL (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MRRREGHGTDSEMGQGPVRESQSSDPPALQFRISEYKPLNMAGVEQPPTPELRQEGVTEYEDGGAPAGDGEAGPQQAEDHPQNPPEDPNQDPPEDDSTCQCQACGPHQAAGPDLGSSNDGCPQLFQERSVIVENSSGSTSASELLKPMKKRKRREYQSPSEEESEPEAMEKQEEGKDPEGQPTASTPESEEWSSSQPATGEKKECWSWESYLEEQKAITAPVSLFQDSQAVTHNKNGFKLGMKLEGIDPQHPSMYFILTVAEVCGYRLRLHFDGYSECHDFWVNANSPDIHPAGWFEKTGHKLQPPKGYKEEEFSWSQYLRSTRAQAAPKHLFVSQSHSPPPLGFQVGMKLEAVDRMNPSLVCVASVTDVVDSRFLVHFDNWDDTYDYWCDPSSPYIHPVGWCQKQGKPLTPPQDYPDPDNFCWEKYLEETGASAVPTWAFKVRPPHSFLVNMKLEAVDRRNPALIRVASVEDVEDHRIKIHFDGWSHGYDFWIDADHPDIHPAGWCSKTGHPLQPPLGPREPSSASPGGCPPLSYRSLPHTRTSKYSFHHRKCPTPGCDGSGHVTGKFTAHHCLSGCPLAERNQSRLKAELSDSEASARKKNLSGFSPRKKPRHHGRIGRPPKYRKIPQEDFQTLTPDVVHQSLFMSALSAHPDRSLSVCWEQHCKLLPGVAGISASTVAKWTIDEVFGFVQTLTGCEDQARLFKDEARIVRVTHVSGKTLVWTVAQLGDLVCSDHLQEGKGILETGVHSLLCSLPTHLLAKLSFASDSQY | |
RUO | |
L3MBTL | |
Wheat Germ (in vitro) | |
GST | |
Liquid |