missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human KLRF1 Partial ORF (NP_057607, 69 a.a. - 166 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00051348-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: KCQKGSCSNATQYEDTGDLKVNNGTRRNISNKDLCASRSADQTVLCQSEWLKYQGKCYWFSNEMKSWSDSYVYCLERKSHLLIIHDQLEMAFIQKNLREspecificaciones
NP_057607 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.52kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
KCQKGSCSNATQYEDTGDLKVNNGTRRNISNKDLCASRSADQTVLCQSEWLKYQGKCYWFSNEMKSWSDSYVYCLERKSHLLIIHDQLEMAFIQKNLR | |
RUO | |
KLRF1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
51348 | |
KLRF1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CLEC5C/MGC119907/MGC119908/MGC119909 | |
KLRF1 | |
Recombinant | |
wheat germ expression system |