missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human KIAA0992 Partial ORF (NP_057165, 775 a.a. - 839 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00023022-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Palladin is a component of actin-containing microfilaments that control cell shape, adhesion, and contraction.[supplied by OMIM]
Sequence: MAPFFEMKLKHYKIFEGMPVTFTCRVAGNPKPKIYWFKDGKQISPKSDHYTIQRDLDGTCSLHTTEspecificaciones
NP_057165 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
32.89kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAPFFEMKLKHYKIFEGMPVTFTCRVAGNPKPKIYWFKDGKQISPKSDHYTIQRDLDGTCSLHTT | |
RUO | |
PALLD | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
23022 | |
KIAA0992 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CGI-151/FLJ22190/FLJ38193/FLJ39139/KIAA0992/PNCA1/SIH002 | |
PALLD | |
Recombinant | |
wheat germ expression system |