Learn More
Abnova™ Human KCNV1 Full-length ORF (AAH28739.1, 1 a.a. - 500 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00027012-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Voltage-gated potassium (Kv) channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. This gene encodes a member of the potassium voltage-gated channel subfamily V. This protein is essentially present in the brain, and its role might be to inhibit the function of a particular class of outward rectifier potassium channel types. [provided by RefSeq]
Sequence: MPSSGRALLDSPLDSGSLTSLGSSVFCSEGEGEPLALGDCFTVNVGGSRFVLSQQALSCFPHTRLGKLAVVVASYRRPGALAAVPSPLELCDDANPVDNEYFFDRSSQAFRYVLHYYRTGRLHVMEQLCALSFLQEIQYWGIDELSIDSCCRDRYFRRKELSETLDFKKDTEDQESQHESEQDFSQGPCPTVRQKLWNILEKPGSSTAARIFGVISIIFVVVSIINMALMSAELSWLDLQLLEILEYVCISWFTGEFVLRFLCVRDRCRFLRKVPNIIDLLAILPFYITLLVESLSGSQTTQELENVGRIVQVLRLLRALRMLKLGRHSTGLRSLGMTITQCYEEVGLLLLFLSVGISIFSTVEYFAEQSIPDTTFTSVPCAWWWATTSMTTVGYGDIRPDTTTGKIVAFMCILSGILVLALPIAIINDRFSACYFTLKLKEAAVRQREALKKLTKNIATDSYISVNLRDVYARSIMEMLRLKGRERASTRSSGGDDFWFEspecificaciones
AAH28739.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
82.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HNKA/KCNB3/KV2.3/KV8.1 | |
KCNV1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
27012 | |
KCNV1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MPSSGRALLDSPLDSGSLTSLGSSVFCSEGEGEPLALGDCFTVNVGGSRFVLSQQALSCFPHTRLGKLAVVVASYRRPGALAAVPSPLELCDDANPVDNEYFFDRSSQAFRYVLHYYRTGRLHVMEQLCALSFLQEIQYWGIDELSIDSCCRDRYFRRKELSETLDFKKDTEDQESQHESEQDFSQGPCPTVRQKLWNILEKPGSSTAARIFGVISIIFVVVSIINMALMSAELSWLDLQLLEILEYVCISWFTGEFVLRFLCVRDRCRFLRKVPNIIDLLAILPFYITLLVESLSGSQTTQELENVGRIVQVLRLLRALRMLKLGRHSTGLRSLGMTITQCYEEVGLLLLFLSVGISIFSTVEYFAEQSIPDTTFTSVPCAWWWATTSMTTVGYGDIRPDTTTGKIVAFMCILSGILVLALPIAIINDRFSACYFTLKLKEAAVRQREALKKLTKNIATDSYISVNLRDVYARSIMEMLRLKGRERASTRSSGGDDFWF | |
RUO | |
KCNV1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |