Learn More
Abnova™ Human KCNH2 Partial ORF (AAH01914, 684 a.a. - 773 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00003757-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a voltage-activated potassium channel belonging to the eag family. It shares sequence similarity with the Drosophila ether-a-go-go (eag) gene. Mutations in this gene can cause long QT syndrome type 2 (LQT2). Transcript variants encoding distinct isoforms have been identified. [provided by RefSeq]
Sequence: LGMGWGAGTGLEMPSAASRGASLLNMQSLGLWTWDCLQGHWAPLIHLNSGPPSGAMERSPTWGEAAELWGSHILLPFRIRHKQTLFASLKEspecificaciones
AAH01914 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.53kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
LGMGWGAGTGLEMPSAASRGASLLNMQSLGLWTWDCLQGHWAPLIHLNSGPPSGAMERSPTWGEAAELWGSHILLPFRIRHKQTLFASLK | |
RUO | |
KCNH2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
3757 | |
KCNH2 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ERG1/HERG/HERG1/Kv11.1/LQT2/SQT1 | |
KCNH2 | |
Recombinant | |
wheat germ expression system |