Learn More
Abnova™ Human KCNC3 Partial ORF (NP_004968, 671 a.a. - 757 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00003748-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
The Shaker gene family of Drosophila encodes components of voltage-gated potassium channels and is comprised of four subfamilies. Based on sequence similarity, this gene is similar to one of these subfamilies, namely the Shaw subfamily. The protein encoded by this gene belongs to the delayed rectifier class of channel proteins and is an integral membrane protein that mediates the voltage-dependent potassium ion permeability of excitable membranes. [provided by RefSeq]
Sequence: ALAHEDCPAIDQPAMSPEDKSPITPGSRGRYSRDRACFLLTDYAPSPDGSIRKATGAPPLPPQDWRKPGPPSFLPDLNANAAAWISPEspecificaciones
NP_004968 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.31kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
ALAHEDCPAIDQPAMSPEDKSPITPGSRGRYSRDRACFLLTDYAPSPDGSIRKATGAPPLPPQDWRKPGPPSFLPDLNANAAAWISP | |
RUO | |
KCNC3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
3748 | |
KCNC3 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KSHIIID/KV3.3/SCA13 | |
KCNC3 | |
Recombinant | |
wheat germ expression system |