missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human KALRN Partial ORF (NP_008995, 1187 a.a. - 1287 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00008997-Q01.25ug

Detalles adicionales : Peso : 0.02000kg

 Ver más versiones de este producto

Código de producto. 16055115

  • 508.00€ / 25 microgramos
Fecha estimada de envío: 10-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Huntington's disease (HD), a neurodegenerative disorder characterized by loss of striatal neurons, is caused by an expansion of a polyglutamine tract in the HD protein huntingtin. This gene encodes a protein that interacts with the huntingtin-associated protein 1, which is a huntingtin binding protein that may function in vesicle trafficking. Alternatively spliced transcript variants encoding different isoforms have been described. [provided by RefSeq]

Sequence: SKEETCINVCRVDFSFPHEYFCGVSNAARDFINVILQEDFRRRPTAATCLQHPWLQPHNGSYSKIPLDTSRLACFIERRKHQNDVRPIPNVKSYIVNRVNQ
Especificaciones

Especificaciones

NP_008995
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.85kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
SKEETCINVCRVDFSFPHEYFCGVSNAARDFINVILQEDFRRRPTAATCLQHPWLQPHNGSYSKIPLDTSRLACFIERRKHQNDVRPIPNVKSYIVNRVNQ
RUO
KALRN
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
8997
KALRN (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DUET/FLJ16443/HAPIP/TRAD/duo
KALRN
Recombinant
wheat germ expression system
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales