missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human JPH1 Partial ORF (NP_065698, 501 a.a. - 579 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00056704-Q01.10ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16198796

  • 302.00€ / 10 microgramos


Fecha estimada de envío
26-10-2020
Añadir a la cesta

Descripción

Descripción

Sequence: VADEQVTAIVNKPLMSKAPTKEAGAVVPQSKYSGRHHIPNPSNGELHSQYHGYYVKLNAPQHPPVDVEDGDGSSQSSSA
Especificaciones

Especificaciones

NP_065698
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
34.43kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
VADEQVTAIVNKPLMSKAPTKEAGAVVPQSKYSGRHHIPNPSNGELHSQYHGYYVKLNAPQHPPVDVEDGDGSSQSSSA
RUO
JPH1
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
56704
JPH1 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp762L0313|JP-1|JP1
JPH1
Yes
Videos
Documentos