Learn More
STEMCELL Technologies Human IL-23 ELISA Kit
For detection and measurement of human interleukin 23
1231.20€
Especificaciones
Descripción | Dendritic Cells; Macrophages |
---|---|
Para utilizar con (aplicación) | ELISA |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
17188021
|
STEMCELL Technologies
02016 |
2 Plates |
1231.20€
paquete de 2 |
Fecha estimada de envío: 13-05-2024
Inicie sesión para ver el stock |
|||||
Descripción
- The Human Interleukin 23 (IL-23) ELISA Kit is designed for the quantitative detection and measurement of human IL-23 in biological fluids such as serum, plasma, and cell culture supernatants
- IL-23 is a heterodimeric cytokine composed of a p19 and a p40 subunit; the p40 subunit is shared with IL-12
- IL-23 is secreted by activated macrophages and dendritic cells, and induces the proliferation of memory T cells and the production of IL-17
- The assay is based on the sandwich ELISA method, in which samples are added to ELISA strip plates pre-coated with capture antibodies specific for the cytokine
- The captured cytokine is detected by addition of a biotinylated detection antibody, followed by streptavidin-horseradish peroxidase, which binds the biotinylated antibody
- Addition of the chromogenic enzyme substrate 3,3',5,5' tetramethylbenzidine (TMB) results in a colored product with an intensity directly proportional to the concentration of cytokine in the sample
Especificaciones
Dendritic Cells; Macrophages | |
ELISA |