Learn More
ACROBiosystems Human IL-17 RA / CD217 Protein, His Tag
Marca: ACROBiosystems ILA-H5222-100UG
Detalles adicionales : Peso : 0.00001kg
Descripción
Interleukin 17 receptor A (IL17RA) is also known as cluster of differentiation w217 (CDw217), is a pro-inflammatory cytokine secreted by activated T-lymphocytes, belong to ubiquitous type I membrane glycoprotein, and binds with low affinity to interleukin 17A (IL17A). IL 17R mRNA exhibits a broad tissue distribution, and has been detected in virtually all cells and tissues tested . IL 17RA associates with IL 17RC to form a signaling receptor complex for IL 17 and IL 17F . Ligand and IL 17RA ligation promotes T cell activation and the production of IL - 6, G-CSF, SCF, and multiple pro-inflammatory chemokines. Defects in IL17RA are the cause of familial candidiasis type 5 (CANDF5).
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 34.3 kDa. The protein migrates as 50-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Especificaciones
NP_055154.3 | |
34.3 kDa | |
100 µg | |
Leu 33 - Trp 320 | |
CD217,CDw217,IL-17 RA,IL17R,CANDF5,hIL-17 R | |
Unconjugated | |
PBS, pH7.4 | |
HEK293 | |
Human |
Powder | |
>98% as determined by SDS-PAGE | |
Human IL-17 RA, His Tag (ILA-H5222) is expressed from human 293 cells (HEK293). It contains AA Leu 33 - Trp 320 (Accession # Q96F46-1). | |
-20°C | |
IL-17 RA | |
His Tag | |
Less than 1.0 EU / µg by the LAL method. | |
Bio-Markers and CD Antigens |