missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Human IL-13 R alpha 2 Protein, His Tag

383.89€ - 2519.28€

Specifications

Número de acceso NP_000631.1
Peso molecular 39.0 kDa
Fuente Human IL-13 R alpha 2, His Tag (IL2-H52H5) is expressed from human 293 cells (HEK293). It contains AA Asp 27 - Arg 343 (Accession # Q14627-1).
Inmunógeno Asp 27 - Arg 343
Requisitos de almacenamiento -20°C
View More Specs

Products 2
Product Code Brand Cantidad Price Quantity & Availability  
Product Code Brand Cantidad Price Quantity & Availability  
16487679
missing translation for 'viewDocuments' Promotion Details
ACROBiosystems
IL2-H52H5-1MG
1 mg
2519.28€
1 miligramo
View alternative products
missing translation for 'toSaveUpto' 
»
Fecha estimada de envío: 11-04-2024
para ver el stock
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
16477679
missing translation for 'viewDocuments' Promotion Details
ACROBiosystems
IL2-H52H5-100UG
100 μg
383.89€
100 microgramos
View alternative products
missing translation for 'toSaveUpto' 
»
Fecha estimada de envío: 11-04-2024
para ver el stock
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
Description

Description

Interleukin-13 receptor subunit alpha-2 is also known as IL13Rα2, IL13Ra2 cluster of differentiation 213A2, CD213A2, CT19, IL-13R, IL13BP, and is a membrane bound protein that in humans is encoded by the IL13RA2 gene. IL13Rα2 is closely related to IL13Rα1, a subunit of the interleukin-13 receptor complex. This protein binds IL13 with high affinity, but lacks any significant cytoplasmic domain, and does not appear to function as a signal mediator. It is, however able to regulate the effects of both IL13 and IL4, despite the fact it is unable to bind directly to the latter. It is also reported to play a role in the internalization of IL13. IL13Rα2 is a component of the cell surface receptors, however, the majority exists in intracellular pools and in soluble form, and thus plays an opposite role as a potent IL13 antagonist compared with IL13Rα1. It also functions as an inhibitor of IL4-dependent pathway probably through the physical interaction between the short intracellular domain of and cytoplasmic domain of IL13Rα2 and the IL4Rα chain. In spite of the failed STAT signaling function, IL13Rα2 dose induce TGF-beta production and fibrosis. Additionally, IL13Rα2has been reported to be abundantly and specifically overexpressed in glioblastoma multiforme.

This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 39.0 kDa. The protein migrates as 45-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Specifications

Specifications

NP_000631.1
Human IL-13 R alpha 2, His Tag (IL2-H52H5) is expressed from human 293 cells (HEK293). It contains AA Asp 27 - Arg 343 (Accession # Q14627-1).
-20°C
IL-13 R alpha 2
Human
PBS, pH7.4
HEK293
Bio-Markers and CD Antigens
39.0 kDa
Asp 27 - Arg 343
IL13RA2,CD213A2,CT19,IL-13 R,IL13BP
Unconjugated
His Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>90% as determined by SDS-PAGE
Videos
SDS
Documents

Documents

Certificates