Learn More
STEMCELL Technologies Human IL-13 ELISA Kit
For detection and measurement of human interleukin 13
Marca:  STEMCELL Technologies 02035
Detalles adicionales : Peso : 0.03670kg
 Ver más versiones de este producto
Código de producto. 17188041
  • 4031.52€ / paquete de 10
Fecha estimada de envío: 07-10-2022
para ver el stock
Añadir a la cesta
Descripción
Descripción
  • The Human Interleukin 13 (IL-13) ELISA Kit is designed for the quantitative detection and measurement of IL-13 in biological fluids such as serum, plasma, and cell culture supernatants
  • IL-13 is secreted by Th2 cells, mast cells and NK cells
  • IL-13 is involved in promoting B cell proliferation, induces class switching to IgG4 and IgE, recruits and activates IgE-producing B cells, and activates the JAK/STAT and IRS-1 and IRS-2 pathways
  • The assay is based on the sandwich ELISA method, in which samples are added to ELISA strip plates pre-coated with capture antibodies specific for the cytokine
  • The captured cytokine is detected by addition of a biotinylated detection antibody, followed by streptavidin-horseradish peroxidase (SA-HRP), which binds the biotinylated antibody
  • Addition of the chromogenic enzyme substrate 3,3',5,5' tetramethylbenzidine (TMB) results in a colored product with an intensity directly proportional to the concentration of cytokine in the sample
Especificaciones
Especificaciones
Granulocytes and Subsets; NK Cells; T Cells, CD8+
10 Plates