Learn More
R&D Systems™ Human IL-11 (NP_000632.1) VersaClone cDNA
Specifications
Specifications
| Gene | IL-11 |
| Accession Number | NP_000632.1 |
| Gene ID (Entrez) | 3589 |
| Gene Alias | Adipogenesis inhibitory factor, AGIFoprelvekin, IL-11 Oprelvekin, interleukin 11, interleukin-11 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Product Line | R&D Systems |
| Quantity | 10 μg |