Learn More
ACROBiosystems Human IGFBP-3 Protein, His Tag
Marca: ACROBiosystems IG3-H5229-1mg
Detalles adicionales : Peso : 0.00010kg
Descripción
Insulin-like growth factor-binding protein 3 is also known as IGFBP3, is a protein that, in humans, is encoded by the IGFBP3 gene. IGFBP3 forms a ternary complex of about 140 ∼150 kDa with IGF1 or IGF2 and a glycoprotein insulin-like growth factor acid-labile subunit (ALS), thus alter the interaction of IGFs with their cell surface receptors. IGFBP3 exerts either proapoptotic or growth stimulatory effects depending upon the cellular context. Studies have shown that IGFBP3 can leads to the induction of apoptosis dependent or independent of the IGF-IGF receptor axis, accordingly acts as a negative regulator of tumorigenesis and progressing in certain carcinomas. The highest expression level is found in the nonparanchymal cells of the liver. Expression levels are also higher during extrauterine life and peak during puberty.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 29.6 kDa. The protein migrates as 40-50 kDa under reducing (R) condition (SDS-PAGE).Especificaciones
AAH18962 | |
29.6 kDa | |
1 mg | |
Gly 28 - Lys 291 | |
IGFBP3,BP-53,IBP3 | |
Unconjugated | |
PBS, pH7.4 | |
HEK293 | |
Human |
Powder | |
>95% as determined by SDS-PAGE | |
Human IGFBP-3, His Tag (IG3-H5229) is expressed from human 293 cells (HEK293). It contains AA Gly 28 - Lys 291 (Accession # AAH18962). | |
-20°C | |
IGFBP-3 | |
His Tag | |
Less than 1.0 EU / µg by the LAL method. | |
Bio-Markers and CD Antigens |