missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human IFNA2 Partial ORF (NP_000596.2, 24 a.a. - 133 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00003440-Q01.25ug

Additional Details : Peso : 0.00010kg

 View more versions of this product

Product Code. 16119864

  • 528.00€ / 25 microgramos
Fecha estimada de envío: 10-07-2023
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Description

Description

Sequence: CDLPQTHSLGSRRTLMLLAQMRRISLFSCLKDRHDFGFPQEEFGNQFQKAETIPVLHEMIQQIFNLFSTKDSSAAWDETLLDKFYTELYQQLNDLEACVIQGVGVTETPL
Specifications

Specifications

NP_000596.2
Liquid
3440
IFNA2 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
IFNA/INFA2/MGC125764/MGC125765
IFNA2
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
CDLPQTHSLGSRRTLMLLAQMRRISLFSCLKDRHDFGFPQEEFGNQFQKAETIPVLHEMIQQIFNLFSTKDSSAAWDETLLDKFYTELYQQLNDLEACVIQGVGVTETPL
RUO
IFNA2
Wheat Germ (in vitro)
GST
Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions