Learn More
Abnova™ Human IDH2 Partial ORF (NP_002159, 354 a.a. - 451 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00003418-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Isocitrate dehydrogenases catalyze the oxidative decarboxylation of isocitrate to 2-oxoglutarate. These enzymes belong to two distinct subclasses, one of which utilizes NAD(+) as the electron acceptor and the other NADP(+). Five isocitrate dehydrogenases have been reported: three NAD(+)-dependent isocitrate dehydrogenases, which localize to the mitochondrial matrix, and two NADP(+)-dependent isocitrate dehydrogenases, one of which is mitochondrial and the other predominantly cytosolic. Each NADP(+)-dependent isozyme is a homodimer. The protein encoded by this gene is the NADP(+)-dependent isocitrate dehydrogenase found in the mitochondria. It plays a role in intermediary metabolism and energy production. This protein may tightly associate or interact with the pyruvate dehydrogenase complex. [provided by RefSeq]
Sequence: HYREHQKGRPTSTNPIASIFAWTRGLEHRGKLDGNQDLIRFAQMLEKVCVETVESGAMTKDLAGCIHGLSNVKLNEHFLNTTDFLDTIKSNLDRALGREspecificaciones
NP_002159 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.52kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
HYREHQKGRPTSTNPIASIFAWTRGLEHRGKLDGNQDLIRFAQMLEKVCVETVESGAMTKDLAGCIHGLSNVKLNEHFLNTTDFLDTIKSNLDRALGR | |
RUO | |
IDH2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
3418 | |
IDH2 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ICD-M/IDH/IDHM/IDP/IDPM/mNADP-IDH | |
IDH2 | |
Recombinant | |
wheat germ expression system |