Learn More
Abnova™ Human ICK Partial ORF (AAH35807, 192 a.a. - 292 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00022858-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Eukaryotic protein kinases are enzymes that belong to a very extensive family of proteins which share a conserved catalytic core common with both serine/threonine and tyrosine protein kinases. This gene encodes an intestinal serine/threonine kinase harboring a dual phosphorylation site found in mitogen-activating protein (MAP) kinases. The protein localizes to the intestinal crypt region and is thought to be important in intestinal epithelial cell proliferation and differentiation. Alternative splicing has been observed at this locus and two variants, encoding the same isoform, have been identified. [provided by RefSeq]
Sequence: EVYTLRPLFPGASEIDTIFKICQVLGTPKKTDWPEGYQLSSAMNFRWPQCVPNNLKTLIPNASSEAVQLLRDMLQWDPKKRPTASQVFFHFLVITFISNSEEspecificaciones
AAH35807 | |
Liquid | |
22858 | |
ICK (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KIAA0936/LCK2/MGC46090/MRK | |
ICK | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
EVYTLRPLFPGASEIDTIFKICQVLGTPKKTDWPEGYQLSSAMNFRWPQCVPNNLKTLIPNASSEAVQLLRDMLQWDPKKRPTASQVFFHFLVITFISNSE | |
RUO | |
ICK | |
Wheat Germ (in vitro) | |
GST |