missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human HUWE1 Partial ORF (NP_113584, 4281 a.a. - 4374 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00010075-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: FWRALRSFDQADRAKFLQFVTGTSKVPLQGFAALEGMNGIQKFQIHRDDRSTDRLPSAHTCFNQLDLPAYESFEKLRHMLLLAIQECSEGFGLAEspecificaciones
NP_113584 | |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.08kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
FWRALRSFDQADRAKFLQFVTGTSKVPLQGFAALEGMNGIQKFQIHRDDRSTDRLPSAHTCFNQLDLPAYESFEKLRHMLLLAIQECSEGFGLA | |
RUO | |
HUWE1 | |
Wheat Germ (in vitro) | |
GST |
wheat germ expression system | |
Liquid | |
10075 | |
HUWE1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ARF-BP1|HECTH9|HSPC272|Ib772|KIAA0312|LASU1|MULE|UREB1 | |
HUWE1 | |
Yes |