missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human HPS1 Full-length ORF (NP_872577.1, 1 a.a. - 324 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00003257-P02.25ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16109027

  • 518.16€ / 25 microgramos
Envío estimado: 03-01-2022

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This gene encodes a protein that may play a role in organelle biogenesis associated with melanosomes, platelet dense granules, and lysosomes. The encoded protein is a component of three different protein complexes termed biogenesis of lysosome-related organelles complex (BLOC)-3, BLOC4, and BLOC5. Mutations in this gene are associated with Hermansky-Pudlak syndrome type 1. Multiple transcript variants encoding distinct isoforms have been identified for this gene; the full-length sequences of some of these have not been determined yet. [provided by RefSeq]

Sequence: MKCVLVATEGAEVLFYWTDQEFEESLRLKFGQSENEEEELPALEDQLSTLLAPVIISSMTMLEKLSDTYTCFSTENGNFLYVLHLFGECLFIAINGDHTESEGDLRRKLYVLKYLFEVHFGLVTVDGHLIRKELRPPDLAQRVQLWEHFQSLLWTYSRLREQEQCFAVEALERLIHPQLCELCIEALERHVIQAVNTSPERGGEEALHAFLLVHSKLLAFYSSHSASSLRPADLLALILLVQDLYPSESTAEDDIQPSPRRARSSQNIPVQQAWSPHSTGPTGGSSAETETDSFSLPEEYFTPAPSPGDQSSGEDRRKAGGNNS
Especificaciones

Especificaciones

NP_872577.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
62.9kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HPS/MGC5277
HPS1
Yes
wheat germ expression system
Liquid
3257
HPS1 (Human) Recombinant Protein (P02)
12.5% SDS-PAGE Stained with Coomassie Blue.
MKCVLVATEGAEVLFYWTDQEFEESLRLKFGQSENEEEELPALEDQLSTLLAPVIISSMTMLEKLSDTYTCFSTENGNFLYVLHLFGECLFIAINGDHTESEGDLRRKLYVLKYLFEVHFGLVTVDGHLIRKELRPPDLAQRVQLWEHFQSLLWTYSRLREQEQCFAVEALERLIHPQLCELCIEALERHVIQAVNTSPERGGEEALHAFLLVHSKLLAFYSSHSASSLRPADLLALILLVQDLYPSESTAEDDIQPSPRRARSSQNIPVQQAWSPHSTGPTGGSSAETETDSFSLPEEYFTPAPSPGDQSSGEDRRKAGGNNS
RUO
HPS1
Wheat Germ (in vitro)
GST
Videos
SDS
Documentos
Certificados
Ofertas especiales

Ofertas especiales