Learn More
Abnova™ Human HOXA1 Partial ORF (NP_005513.1, 11 a.a. - 119 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Especificaciones
Número de acceso | NP_005513.1 |
---|---|
Para utilizar con (aplicación) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulación | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
ID de gen (Entrez) | 3198 |
Peso molecular | 37.73kDa |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
16199474
|
Abnova™
H00003198-Q01.10UG |
10 ug |
335.00€
10 microgramos |
Fecha estimada de envío: 30-05-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
16109484
|
Abnova™
H00003198-Q01.25UG |
25 ug |
508.00€
25 microgramos |
Fecha estimada de envío: 30-05-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
Descripción
In vertebrates, the genes encoding the class of transcription factors called homeobox genes are found in clusters named A, B, C, and D on four separate chromosomes. Expression of these proteins is spatially and temporally regulated during embryonic development. This gene is part of the A cluster on chromosome 7 and encodes a DNA-binding transcription factor which may regulate gene expression, morphogenesis, and differentiation. The encoded protein may be involved in the placement of hindbrain segments in the proper location along the anterior-posterior axis during development. Two transcript variants encoding two different isoforms have been found for this gene, with only one of the isoforms containing the homeodomain region. [provided by RefSeq]
Sequence: EYPILSSGDSGTCSARAYPSDHRITTFQSCAVSANSCGGDDRFLVGRGVQIGSPHHHHHHHHHHPQPATYQTSGNLGVSYSHSSCGPSYGSQNFSAPYSPYALNQEADVEspecificaciones
NP_005513.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.73kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BSAS/HOX1/HOX1F/MGC45232 | |
HOXA1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
3198 | |
HOXA1 (Human) Recombinant Protein (Q01) | |
EYPILSSGDSGTCSARAYPSDHRITTFQSCAVSANSCGGDDRFLVGRGVQIGSPHHHHHHHHHHPQPATYQTSGNLGVSYSHSSCGPSYGSQNFSAPYSPYALNQEADV | |
RUO | |
HOXA1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |