Learn More
Abnova™ Human HOOK3 Partial ORF (NP_115786.1, 622 a.a. - 718 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00084376-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Hook proteins are cytosolic coiled-coil proteins that contain conserved N-terminal domains, which attach to microtubules, and more divergent C-terminal domains, which mediate binding to organelles. The Drosophila Hook protein is a component of the endocytic compartment.[supplied by OMIM]
Sequence: QNQGAAPEIQALKNQLQERDRLFHSLEKEYEKTKSQREMEEKYIVSAWYNMGMTLHKKAAEDRLASTGSGQSFLARQRQATSSRRSYPGHVQPATAREspecificaciones
NP_115786.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.41kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
QNQGAAPEIQALKNQLQERDRLFHSLEKEYEKTKSQREMEEKYIVSAWYNMGMTLHKKAAEDRLASTGSGQSFLARQRQATSSRRSYPGHVQPATAR | |
RUO | |
HOOK3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
84376 | |
HOOK3 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ31058/HK3 | |
HOOK3 | |
Recombinant | |
wheat germ expression system |