missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human HMGB3 Full-length ORF (NP_005333.2, 1 a.a. - 200 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00003149-P01.10ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16196221

  • 335.00€ / 10 microgramos
Fecha estimada de envío: 04-06-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

HMGB3 belongs to the high mobility group (HMG) protein superfamily. Like HMG1 (MIM 163905) and HMG2 (MIM 163906), HMGB3 contains DNA-binding HMG box domains and is classified into the HMG box subfamily. Members of the HMG box subfamily are thought to play a fundamental role in DNA replication, nucleosome assembly and transcription (Wilke et al., 1997 [PubMed 9370291]; Nemeth et al., 2006 [PubMed 16945912]).[supplied by OMIM]

Sequence: MAKGDPKKPKGKMSAYAFFVQTCREEHKKKNPEVPVNFAEFSKKCSERWKTMSGKEKSKFDEMAKADKVRYDREMKDYGPAKGGKKKKDPNAPKRPPSGFFLFCSEFRPKIKSTNPGISIGDVAKKLGEMWNNLNDSEKQPYITKAAKLKEKYEKDVADYKSKGKFDGAKGPAKVARKKVEEEDEEEEEEEEEEEEEEDE
Especificaciones

Especificaciones

NP_005333.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
49.4kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HMG2A/HMG4/MGC90319
HMGB3
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
3149
HMGB3 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAKGDPKKPKGKMSAYAFFVQTCREEHKKKNPEVPVNFAEFSKKCSERWKTMSGKEKSKFDEMAKADKVRYDREMKDYGPAKGGKKKKDPNAPKRPPSGFFLFCSEFRPKIKSTNPGISIGDVAKKLGEMWNNLNDSEKQPYITKAAKLKEKYEKDVADYKSKGKFDGAKGPAKVARKKVEEEDEEEEEEEEEEEEEEDE
RUO
HMGB3
Wheat Germ (in vitro)
GST
Liquid
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales