Learn More
Abnova™ Human HMGB3 Full-length ORF (NP_005333.2, 1 a.a. - 200 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00003149-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
HMGB3 belongs to the high mobility group (HMG) protein superfamily. Like HMG1 (MIM 163905) and HMG2 (MIM 163906), HMGB3 contains DNA-binding HMG box domains and is classified into the HMG box subfamily. Members of the HMG box subfamily are thought to play a fundamental role in DNA replication, nucleosome assembly and transcription (Wilke et al., 1997 [PubMed 9370291]; Nemeth et al., 2006 [PubMed 16945912]).[supplied by OMIM]
Sequence: MAKGDPKKPKGKMSAYAFFVQTCREEHKKKNPEVPVNFAEFSKKCSERWKTMSGKEKSKFDEMAKADKVRYDREMKDYGPAKGGKKKKDPNAPKRPPSGFFLFCSEFRPKIKSTNPGISIGDVAKKLGEMWNNLNDSEKQPYITKAAKLKEKYEKDVADYKSKGKFDGAKGPAKVARKKVEEEDEEEEEEEEEEEEEEDEEspecificaciones
NP_005333.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
49.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HMG2A/HMG4/MGC90319 | |
HMGB3 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
3149 | |
HMGB3 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAKGDPKKPKGKMSAYAFFVQTCREEHKKKNPEVPVNFAEFSKKCSERWKTMSGKEKSKFDEMAKADKVRYDREMKDYGPAKGGKKKKDPNAPKRPPSGFFLFCSEFRPKIKSTNPGISIGDVAKKLGEMWNNLNDSEKQPYITKAAKLKEKYEKDVADYKSKGKFDGAKGPAKVARKKVEEEDEEEEEEEEEEEEEEDE | |
RUO | |
HMGB3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |