Learn More
ACROBiosystems Human Her2 / ErbB2 (23-450) Protein, Fc Tag
en la oferta
Especificaciones
Número de acceso | NP_004439.2 |
---|---|
Peso molecular | 73.8 kDa |
Fuente | Human Her2 (23-450), Fc Tag (HE2-H525x) is expressed from human 293 cells (HEK293). It contains AA Thr 23 - Ile 450 (Accession # P04626-1). |
Inmunógeno | Thr 23 - Ile 450 |
Requisitos de almacenamiento | -20°C |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
15896927
|
ACROBiosystems
HE2-H525X-25UG |
25 μg |
en la oferta
25 microgramos |
N/A | Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
Este artículo requiere una actualización de precios antes de ser solicitado. Solicite el precio de este artículo aquí. |
|||||||||
16427629
|
ACROBiosystems
HE2-H525X-1MG |
1 mg |
en la oferta
1 miligramo |
N/A | Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
Este artículo requiere una actualización de precios antes de ser solicitado. Solicite el precio de este artículo aquí. |
|||||||||
Description
Human Epidermal growth factor Receptor 2 (HER2) is also called ERBB2, HER-2,HER-2 /neu, NEU, NGL,TKR1 and c-erb B2,and is a protein giving higher aggressiveness in breast cancers. It is a member of the ErbB protein family, more commonly known as the epidermal growth factor receptor family. HER2 is a cell membrane surface-bound receptor tyrosine kinase and is normally involved in the signal transduction pathways leading to cell growth and differentiation. HER2 is thought to be an orphan receptor, with none of the EGF family of ligands able to activate it. Approximately 30% of breast cancers have an amplification of the HER2 gene or overexpression of its protein product. Overexpression of this receptor in breast cancer is associated with increased disease recurrence and worse prognosis. HER2 appears to play roles in development, cancer, communication at the neuromuscular junction and regulation of cell growth and differentiation .
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 73.8 kDa. The protein migrates as 95-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifications
NP_004439.2 | |
Human Her2 (23-450), Fc Tag (HE2-H525x) is expressed from human 293 cells (HEK293). It contains AA Thr 23 - Ile 450 (Accession # P04626-1). | |
-20°C | |
Her2 | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Bio-Markers and CD Antigens |
73.8 kDa | |
Thr 23 - Ile 450 | |
ERBB2,CD340,HER-2/neu,HER2,MLN19,NEU,NGL,TKR1 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |