missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Human Hepatitis D Virus Antigen Antibody (IgG) ELISA Kit (Colorimetric)

Colorimetric ELISA Kit is useful in study of hepatitis infection

Marca:  Novus Biologicals™ NBP2-60047-1KIT

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18214371

  • 831.51€ / 1 set
Fecha estimada de envío: 14-08-2024
para ver el stock

Descripción

Descripción

The Human Hepatitis D Virus Antigen Antibody (IgG) ELISA Kit (Colorimetric) measures human Hepatitis D Virus Antigen Antibody (IgG) in serum, plasma.
Especificaciones

Especificaciones

Qualitative
Plasma, Serum
Human
Coated assay plate, Negative Control, Positive Control, Sample Diluents, HRP- conjugate, Wash Buffer (30X concentrate), Substrate A, Substrate B, Stop Solution, Adhesive Strip (For 96 wells), Instruction manual
RUO
50 to 100 μL
Hepatitis D Virus Antigen Antibody (IgG)
ELISA Kit (Colorimetric)
Detection of human HDV antibody(IgG)
ELISA
1 Kit
Cell Biology
Store the unopened product at 2° to 8°C. Protect from light. Do not use past expiration date.
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales

For Research Use Only