Learn More
Abnova™ Human HECA Partial ORF (NP_057301, 434 a.a. - 543 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00051696-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes the homolog of the Drosophila headcase protein, a highly basic, cytoplasmic protein that regulates the re-entry of imaginal cells into the mitotic cycle during adult morphogenesis. In Drosophila, the encoded protein also inhibits terminal branching of neighboring cells during tracheal development. [provided by RefSeq]
Sequence: CHLQGRLMHLYAVCVDCLEGVHKIICIKCKSRWDGSWHQLGTMYTYDILAASPCCQARLNCKHCGKPVIDVRIGMQYFSEYSNVQQCPHCGNLDYHFVKPFSSFKVLEAYEspecificaciones
NP_057301 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
CHLQGRLMHLYAVCVDCLEGVHKIICIKCKSRWDGSWHQLGTMYTYDILAASPCCQARLNCKHCGKPVIDVRIGMQYFSEYSNVQQCPHCGNLDYHFVKPFSSFKVLEAY | |
RUO | |
HECA | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
51696 | |
HECA (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HDC/HDCL/HHDC/dJ225E12.1 | |
HECA | |
Recombinant | |
wheat germ expression system |