missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human HDDC2 Full-length ORF (AAH66332.1, 1 a.a. - 204 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00051020-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: MASVSSATFSGHGARSLLQFLRLVGQLKRVPRTGWVYRNVQRPESVSDHMYRMAVMAMVIKDDRLNKDRCVRLALVHDMAECIVGDIAPADNIPKEEKHRREEEAMKQITQLLPEDLRKELYELWEEYETRSSAEAKFVKQLDQCEMILQASEYEDLEHKPGRLQDFYDSTAGKFNHPEIVQLVSELEAERSTNIAAAASEPHSEspecificaciones
AAH66332.1 | |
Liquid | |
51020 | |
HDDC2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MASVSSATFSGHGARSLLQFLRLVGQLKRVPRTGWVYRNVQRPESVSDHMYRMAVMAMVIKDDRLNKDRCVRLALVHDMAECIVGDIAPADNIPKEEKHRREEEAMKQITQLLPEDLRKELYELWEEYETRSSAEAKFVKQLDQCEMILQASEYEDLEHKPGRLQDFYDSTAGKFNHPEIVQLVSELEAERSTNIAAAASEPHS | |
RUO | |
HDDC2 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
49.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C6orf74/CGI-130/MGC87330/NS5ATP2/dJ167O5.2 | |
HDDC2 | |
Yes | |
wheat germ expression system |