missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human HBQ1 Partial ORF (NP_005322, 1 a.a. - 78 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00003049-Q01.25ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16109194

  • 508.00€ / 25 microgramos
Fecha estimada de envío: 10-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Theta-globin mRNA is found in human fetal erythroid tissue but not in adult erythroid or other nonerythroid tissue. The theta-1 gene may be expressed very early in embryonic life, perhaps sometime before 5 weeks. Theta-1 is a member of the human alpha-globin gene cluster that involves five functional genes and two pseudogenes. The order of genes is: 5' - zeta - pseudozeta - mu - pseudoalpha-2 -pseudoalpha-1 - alpha-2 - alpha-1 - theta-1 - 3'. Research supports a transcriptionally active role for the gene and a functional role for the peptide in specific cells, possibly those of early erythroid tissue. [provided by RefSeq]

Sequence: MALSAEDRALVRALWKKLGSNVGVYTTEALERTFLAFPATKTYFSHLDLSPGSSQVRAHGQKVADALSLAVERLDDLP
Especificaciones

Especificaciones

NP_005322
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
34.32kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MALSAEDRALVRALWKKLGSNVGVYTTEALERTFLAFPATKTYFSHLDLSPGSSQVRAHGQKVADALSLAVERLDDLP
RUO
HBQ1
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
3049
HBQ1 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HBQ1
Wheat Germ (in vitro)
GST
Liquid
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales