missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human GRPEL2 Full-length ORF (NP_689620.2, 1 a.a. - 225 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00134266-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: MAVRSLWAGRLRVQRLLAWSAAWESKGWPLPFSTATQRTAGEDCRSEDPPDELGPPLAERALRVKAVKLEKEVQDLTVRYQRAIADCENIRRRTQRCVEDAKIFGIQSFCKDLVEVADILEKTTECISEESEPEDQKLTLEKVFRGLLLLEAKLKSVFAKHGLEKLTPIGDKYDPHEHELICHVPAGVGVQPGTVALVRQDGYKLHGRTIRLARVEVAVESQRRLEspecificaciones
NP_689620.2 | |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
51.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp451C205|FLJ23713|FLJ33918|Mt-GrpE#2 | |
GRPEL2 | |
Yes |
wheat germ expression system | |
Liquid | |
134266 | |
GRPEL2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAVRSLWAGRLRVQRLLAWSAAWESKGWPLPFSTATQRTAGEDCRSEDPPDELGPPLAERALRVKAVKLEKEVQDLTVRYQRAIADCENIRRRTQRCVEDAKIFGIQSFCKDLVEVADILEKTTECISEESEPEDQKLTLEKVFRGLLLLEAKLKSVFAKHGLEKLTPIGDKYDPHEHELICHVPAGVGVQPGTVALVRQDGYKLHGRTIRLARVEVAVESQRRL | |
RUO | |
GRPEL2 | |
Wheat Germ (in vitro) | |
GST |