missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human GPR92 Partial ORF (NP_065133.1, 301 a.a. - 370 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

335.00€ - 508.00€

Especificaciones

Número de acceso NP_065133.1
Para utilizar con (aplicación) Antibody Production, ELISA, Protein Array, Western Blot
Formulación 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
ID de gen (Entrez) 57121
Peso molecular 33.44kDa
Ver más especificaciones

Productos 2
Product Code Brand Cantidad Price Quantity & Availability  
Product Code Brand Cantidad Price Quantity & Availability  
16109016
missing translation for 'viewDocuments' Promotion Details
Abnova™
H00057121-Q01.10UG
10 ug
335.00€
10 microgramos
View alternative products
missing translation for 'toSaveUpto' 
»
Fecha estimada de envío: 10-05-2024
para ver el stock
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
16119016
missing translation for 'viewDocuments' Promotion Details
Abnova™
H00057121-Q01.25UG
25 ug
508.00€
25 microgramos
View alternative products
missing translation for 'toSaveUpto' 
»
Fecha estimada de envío: 10-05-2024
para ver el stock
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
Description

Description

This gene encodes a member of the rhodopsin class of G protein-coupled transmembrane receptors. This protein transmits extracellular signals from lysophosphatidic acid to cells through heterotrimeric G proteins and mediates numerous cellular processes. Many G protein receptors serve as targets for pharmaceutical drugs. Transcript variants of this gene have been described.[provided by RefSeq]

Sequence: AEGFRNTLRGLGTPHRARTSATNGTRAALAQSERSAVTTDATRPDAASQGLLRPSDSHSLSSFTQCPQDS
Specifications

Specifications

NP_065133.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
33.44kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
GPR92/GPR93/KPG_010/LPA5
LPAR5
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
57121
GPR92 (Human) Recombinant Protein (Q01)
AEGFRNTLRGLGTPHRARTSATNGTRAALAQSERSAVTTDATRPDAASQGLLRPSDSHSLSSFTQCPQDS
RUO
LPAR5
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Certificates