missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human GPR172B Full-length ORF (NP_060456.2, 1 a.a. - 448 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00055065-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
GPCR41 (MIM 607882) and GPCR42 act as receptors for porcine endogenous retrovirus subgroup A (PERV-A).[supplied by OMIM]
Sequence: MAAPTLGRLVLTHLLVALFGMGSWAAVNGIWVELPVVVKDLPEGWSLPSYLSVVVALGNLGLLVVTLWRRLAPGKGEQVPIQVVQVLSVVGTALLAPLWHHVAPVAGQLHSVAFLTLALVLAMACCTSNVTFLPFLSHLPPPFLRSFFLGQGLSALLPCVLALVQGVGRLECPPAPTNGTSGPPLDFPERFPASTFFWALTALLVTSAAAFRGLLLLLPSLPSVTTGGSGPELQLGSPGAEEEEKEEEEALPLQEPPSQAAGTIPGPDPEVHQLFSAHGAFLLGLMAFTSAVTNGVLPSVQSFSCLPYGRLAYHLAVVLGSAANPLACFLAMGVLCRSLAGLVGLSLLGMLFGAYLMALAILSPCPPLVGTTAGVVLVVLSWVLCLCVFSYVKVAASSLLHGGGRPALLAAGVAIQVGSLLGAGAMFPPTSIYHVFQSRKDCVDPCGPEspecificaciones
NP_060456.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
72.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ10060/GPCR/GPCR42/PAR2/RFT1 | |
GPR172B | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
55065 | |
GPR172B (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAAPTLGRLVLTHLLVALFGMGSWAAVNGIWVELPVVVKDLPEGWSLPSYLSVVVALGNLGLLVVTLWRRLAPGKGEQVPIQVVQVLSVVGTALLAPLWHHVAPVAGQLHSVAFLTLALVLAMACCTSNVTFLPFLSHLPPPFLRSFFLGQGLSALLPCVLALVQGVGRLECPPAPTNGTSGPPLDFPERFPASTFFWALTALLVTSAAAFRGLLLLLPSLPSVTTGGSGPELQLGSPGAEEEEKEEEEALPLQEPPSQAAGTIPGPDPEVHQLFSAHGAFLLGLMAFTSAVTNGVLPSVQSFSCLPYGRLAYHLAVVLGSAANPLACFLAMGVLCRSLAGLVGLSLLGMLFGAYLMALAILSPCPPLVGTTAGVVLVVLSWVLCLCVFSYVKVAASSLLHGGGRPALLAAGVAIQVGSLLGAGAMFPPTSIYHVFQSRKDCVDPCGP | |
RUO | |
GPR172B | |
Wheat Germ (in vitro) | |
GST | |
Liquid |