Learn More
Abnova™ Human GPR161 Partial ORF (NP_722561.1, 362 a.a. - 460 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00023432-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Upon ligand binding, G protein-coupled receptors, such as GPR161, activate cytoplasmic G proteins (see GNAS, MIM 139320), allowing the receptors to transduce extracellular signals across the plasma membrane into the cell. Phosphorylation of the receptor attenuates signaling (Matteson et al., 2008 [PubMed 18250320]).[supplied by OMIM]
Sequence: SNRITDLGLSPHLTALMAGGQPLGHSSSTGDTGFSCSQDSGTDMMLLEDYTSDDNPPSHCTCPPKRRSSVTFEDEVEQIKEAAKNSILHVKAEVHKSLDEspecificaciones
NP_722561.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
SNRITDLGLSPHLTALMAGGQPLGHSSSTGDTGFSCSQDSGTDMMLLEDYTSDDNPPSHCTCPPKRRSSVTFEDEVEQIKEAAKNSILHVKAEVHKSLD | |
RUO | |
GPR161 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
23432 | |
GPR161 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ33952/RE2 | |
GPR161 | |
Recombinant | |
wheat germ expression system |